Skip to Content
Merck
All Photos(1)

Documents

CLS431532

Corning® square PET storage bottles

square clear PET, sterile, capacity 500 mL, screw cap, case of 24

Synonym(s):

Corning bottles, PET bottles, screw cap bottles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

screw cap
square clear PET
wide-mouth bottle

description

individually wrapped

sterility

sterile
sterile

feature

cap
graduations

packaging

case of 24

manufacturer/tradename

Corning 431532

bottle neck diam.

37 mm

capacity

500 mL

fitting

45

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® square PET storage bottles are ideal for storing buffers, media, biological, and other aqueous solutions.
  • The polyethylene terephthalate (PET) resin makes the bottle lightweight, break resistant and transparent as glass.
  • Large opening enables easier access for liquid transfer.
  • Features linerless, one-piece autoclavable orange GL45 threaded polypropylene plug seal cap.
  • Screened white enamel graduations are easier to read.
  • Large white marking spot for easy sample identification.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  5. What is the thickness of Corning® square PET storage bottles?

    The thickness for all these PET bottles is 0.95 mm.

  6. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service