CLS431487
Corning® Spin-X® UF concentrators
Spin-X UF 20 for samples up to 20mL; 5K MWCO
Synonym(s):
centrifuge columns, desalting columns
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
polycarbonate housing
polyethersulfone membrane
description
white, polypropylene screw cap
sterility
non-sterile
packaging
case of 12 ea
manufacturer/tradename
Corning 431487
H × diam.
116 mm × 30 mm
membrane surface area
6 cm2
tube size
50 mL
working volume
14 mL , fixed angle rotor
pore size
5 kDa MWCO
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Corning Spin-X UF Concentrators:
Summary:
Note: Spin-X UF 500 concentrators require fixed angle rotors. Minimum rotor angle is 40º
- Corning® Spin-X® UF centrifugal concentrators offer a simple, one step procedure for concentrating or desalting proteins and other biomolecules with 90% or better recovery
- The vertical membrane design and thin channel filtration chamber minimizes membrane fouling and provides fast, high speed concentrating, even with particle laden solutions
- Integrated dead stop design eliminates the risk of spinning to dryness; no respinning necessary
- Choice of three sizes for greater flexibility: Spin-X UF 500 for samples up to 500 μL, Spin-X UF 6 for samples up to 6mL, Spin-X UF 20 for samples up to 20 mL (14 mL if using fixed angle rotors)
- Low binding polyethersulfone (PES) membranes are available with five molecular weight cut-offs (MWCO): 5K, 10K, 30K, 50K and 100K to meet all your concentrating needs. Note: Choose a MWCO one-half to one-third smaller than the protein to be concentrated
- The MWCO and graduations are printed right on the side of the concentrator tube to avoid confusion
Summary:
- Spin-X UF 500 for samples up to 500 μL
- Spin-X UF 6 for samples up to 6 mL
- Spin-X UF 20 for samples up to 20 mL (14 mL if using fixed angle rotors)
Note: Spin-X UF 500 concentrators require fixed angle rotors. Minimum rotor angle is 40º
Note: sample volume of 14mL if using fixed angle rotors
Application
Key Spin-X UF Concentrator Applications:
- Concentration, desalting of proteins, enzymes, monoclonal antibodies, immunoglobulins
- Removal of labeled amino acids and nucleotides
- Deproteinization of samples
- HPLC sample preparation
- Recovery of biomolecules from cell culture supernatants, lysates
- Concentrating virus from cell culture medium
Legal Information
Corning is a registered trademark of Corning, Inc.
SPIN-X is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service