Skip to Content
Merck
All Photos(1)

Key Documents

CLS3936

Corning® 1536 well plates

non-treated (medium bind), 50/cs, round bottom black polystyrene wells, non-sterile, lid: no

Synonym(s):

1536 multiwell plate, 1536 well microplate, 1536 well microtiter plate, 1536 well plate, HTS plate, TC-treated plate, immunoassay plate, tissue-culture treated plate

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
€1,590.00

€1,590.00


Please contact Customer Service for Availability


Select a Size

Change View
50 EA
€1,590.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black plate
polystyrene plate
round bottom black polystyrene wells
size:
1536 wells

€1,590.00


Please contact Customer Service for Availability

material

black plate
polystyrene plate
round bottom black polystyrene wells

sterility

non-sterile

feature

lid: no
skirt

packaging

pack of 10
case of 50

manufacturer/tradename

Corning 3936

maximum volume

10 μL

size

1536 wells

working volume

8 μL

color

black

suitability

suitable for (fluorescent; luminescent assays)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning 1536 Well Plates

  • 1536 well polystyrene plates
  • Low background fluorescence, minimal light scatter and reduced crosstalk
  • Excellent signal dynamic range due to low background fluorescence and enhanced signal intensity
  • Opaque walls to prevent well-to-well crosstalk
  • Flat well bottoms possess superior flatness -minimizes variability within a plate and between plates
  • Round well bottoms for reduced air entrapment and improved CV′s and Z factor
  • Raised well bottoms for high sensitivity
  • Flood reservior on four sides to reduce instrument contamination
  • Suitable for fluorescent and luminescent assays
  • Black wall plates have low background fluorescence, minimal light scatter and reduced crosstalk
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200 ng IgG/cm2
  • Recommended working volume of up to 8 μL
  • Conforms to standard microplate footprint and dimensions

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service