Skip to Content
Merck
All Photos(2)

Key Documents

CLS3916

Corning® 96 Well Solid Polystyrene Microplate

flat bottom, black polystyrene, Tissue Culture (TC)-treated surface, bag of 20, sterile, lid

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€703.00

€703.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EA
€703.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
black bottom
black polystyrene
flat bottom
size:
96 wells

€703.00


Please contact Customer Service for Availability

material

black bottom
black polystyrene
flat bottom

sterility

sterile

feature

lid
skirt

packaging

bag of 20

manufacturer/tradename

Corning 3916

size

96 wells

surface area

0.32 cm2 , cell growth area

cell growth area

cell growth area

well volume

360 μL

working volume

75-200 μL

color

black

suitability

suitable for (cell culture)

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Solid Polystyrene Microplate Black wall plates display low background fluorescence, reduced crosstalk and minimal scattering.
  • Opaque black polystyrene walls with minimal crosstalk and background
  • luminescence.
  • Treated for optimal cell attachment for working volumes of 75 to 200 μL.
  • Non-reversible lids with condensation ring to reduce contamination.
  • Sterilized by gamma radiation and nonpyrogenic.
  • Alphanumeric codes for individual wells.
  • Hydrophilic and negatively charged surface optimized for cell attachment and growth.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service