Skip to Content
Merck
All Photos(1)

Key Documents

CLS3847

Corning® 384 well microplate

clear polystyrene, flat bottom, non-sterile, lid, pack of 20 ea

Synonym(s):

384 multiwell plates

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€3,800.00

€3,800.00


Please contact Customer Service for Availability


Select a Size

Change View
100 EA
€3,800.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.13
material:
clear polystyrene
flat bottom
size:
384 wells

€3,800.00


Please contact Customer Service for Availability

material

clear polystyrene
flat bottom

sterility

non-sterile

feature

lid

packaging

case of 100 ea
pack of 20 ea

manufacturer/tradename

Corning 3847

size

384 wells

color

clear

suitability

suitable for (cell cuture)

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service