CLS3824
Corning® 384 well microplate, low volume
NBS™ (non-binding surface), white polystyrene, flat bottom, non-sterile, lid: no, pack of 10
Synonym(s):
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates
Select a Size
€636.00
Select a Size
About This Item
white bottom
white polystyrene
skirt
€636.00
Recommended Products
material
flat bottom
white bottom
white polystyrene
sterility
non-sterile
feature
lid: no
skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3824
size
384 well
wells
384 wells , low volume
working volume
5-40 μL
color
white
binding type
NBS™ (non-binding surface)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Round well design with a solid white flat bottom.
- 2.8 mm distance between well bottoms to plate bottom.
- Ideal for miniaturized 384 well fluorescent homogeneous assays.
- Nonbinding surface (NBS™) is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions.
- Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise
Note: Suitable for 384 well fluorescent homogeneous assays
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service