Skip to Content
Merck
All Photos(1)

Key Documents

CLS3725BC

Corning® 1536 well plate, white, FB, PS, NT

Synonym(s):

1536 multiwell plate

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
€1,310.00

€1,310.00


Please contact Customer Service for Availability


Select a Size

Change View
50 EA
€1,310.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
flat wells
polystyrene plate
square wells
white bottom
white wells
size:
1536 wells

€1,310.00


Please contact Customer Service for Availability

material

flat wells
polystyrene plate
square wells
white bottom
white wells

sterility

non-sterile

feature

lid: no
skirt

packaging

pack of 10 ea
case of 50 ea

manufacturer/tradename

Corning 3725BC

availability

available only in South Korea

size

1536 wells

well volume

12.5 μL

well working volume

1-8 μL

color

white

suitability

suitable for (cell cuture)

binding type

non-treated surface

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service