Skip to Content
Merck
All Photos(1)

Key Documents

CLS3550

Corning® 96 well protein crystallization plates

COC, non-treated, 3:1 1uL conical flat bottom crystal cup wells, clear, nonsterile

Synonym(s):

96 well protein crystallization plate

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
€847.00

€847.00


Estimated to ship on26 June 2025Details



Select a Size

Change View
50 EA
€847.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
clear plate
conical bottom clear wells
cyclic olefin copolymer (COC)
size:
96 wells

€847.00


Estimated to ship on26 June 2025Details


material

clear plate
conical bottom clear wells
cyclic olefin copolymer (COC)

sterility

non-sterile

feature

lid: no
(protein wells: 3/reservoir well)
plate format: 96 well

packaging

pack of 10 ea
case of 50 ea

manufacturer/tradename

Corning 3550

reservoir volume

100 μL

size

96 wells

well volume

1 μL , protein well volume

color

clear

suitability

suitable for (sitting drop vapor diffusion high-throughput (HT) protein crystallization)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Each well contains three 1 μL conical flat bottom Crystal Cup protein wells and one 100 μL reagent reservoir
  • Crystal Cup well design facilitates centering and collecting protein crystals
  • Low volume 100 μL reagent reservoir saves on reagent costs
  • Designed for sitting drop vapor diffusion high throughput protein crystallization
  • Manufactured from an advanced Cyclic Olefin Copolymer (COC) with high resistance to commonly used solvents, including acetone, acetic acid, butanone, ethanol, isopropanol, methanol, DMSO, nitric acid (65%), sulfuric acid (40%), hydrochloric acid (36%), and ammonia solution (33%) and improved transparency
  • Allows protein crystals to be easily viewed under polarized light with minimal background interference
  • Low water absorption of the polymer prevents loss of protein drop volume
  • Reservoir numbers are embossed on each individual well for easy identification
The Corning 96 well protein crystallization plate features large reservoir wells with corresponding protein wells. It is made of an advanced optical polymer (cyclic olefin copolymer (COC)) superior to polypropylene and polystyrene, which has higher optical clarity and greater resistance to organic solvents. The plate can be used for the sitting-drop method for high-throughput protein crystallization, and is compatible with manual pipettors and robotic operations.

The large reservoir well has a maximum volume of 100 μL, with a recommended working volume between 25 to 755 μL for screen reagents. The small protein well has a volume range of 1-4 μL.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service