Skip to Content
Merck
All Photos(2)

Key Documents

CLS354639

Corning® Vented Screw Cap

for use with Falcon® 175cm Flasks, sterile

Synonym(s):

Vented Screw Cap

Sign Into View Organizational & Contract Pricing

Select a Size

50 EACH
€111.00

€111.00


Please contact Customer Service for Availability


Select a Size

Change View
50 EACH
€111.00

About This Item

UNSPSC Code:
41122104
NACRES:
NB.13

€111.00


Please contact Customer Service for Availability

material

PE flask

sterility

sterile

packaging

pack of 50 ea (Individually wrapped)

manufacturer/tradename

Corning 354639

compatibility

for use with Falcon® 175cm Flasks

Looking for similar products? Visit Product Comparison Guide

General description

Vented Screw Cap for Falcon® 175cm2 Flasks, Bulk, Sterile, 50/Case

Application

Cell Culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service