Skip to Content
Merck
All Photos(2)

Key Documents

CLS354239

Corning® Ultrapure Laminin

from mouse, pack of 1 mg

Synonym(s):

extracellular matrix

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€237.00

€237.00


Please contact Customer Service for Availability


Select a Size

Change View
1 EACH
€237.00

About This Item

UNSPSC Code:
41116155
NACRES:
NA.7

€237.00


Please contact Customer Service for Availability

biological source

mouse

form

liquid

packaging

pack of 1 mg

manufacturer/tradename

Corning 354239

technique(s)

cell attachment: suitable
cell differentiation: suitable

Related Categories

General description

Corning Ultrapure Laminin, mouse, 1mg, is entactic free and is used as a thin coating on tissue-culture surfaces or as a soluble additive to culture medium. Laminin is a major component of basement membranes. It has numerous biological activities including promotion of cell adhesion, migration, chemotaxis, growth, and differentiation, including neurite outgrowth.
Quality
Purity > 95% by SDS-PAGE
Tested for ability to initiate neurite outgrowth of NG-108 human neuroblastoma cells
Found negative for bacteria, fungi, and mycoplasma

Source
Engelbreth-Holm-Swarm (EHS) mouse tumor

Molecular Weight
900 kD (unreduced)

Preparation and Storage
Stable for at least three months at -70°C

Handling
Recommended concentration - 1-10 μg/cm² of growth surface depending on cell type

Formulation
Frozen in 0.05 M Tris-HCl, 0.15 M NaCl, pH 7.4

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service