CLS353810
Corning® Primaria Cell Culture Flask
capacity 150 mL, straight neck, cap, vented
Synonym(s):
Corning container
Select a Size
€477.00
Select a Size
About This Item
€477.00
Recommended Products
material
polystyrene
feature
cap vented
manufacturer/tradename
Corning 353810
technique(s)
cell culture | mammalian: suitable
capacity
150 mL
Looking for similar products? Visit Product Comparison Guide
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service