CLS3480
Corning® NETWELL® 6 well plates
clear , polyester membrane, round wells, clear wells, polyester plate, sterile, pkg of 48 (8 x 6 well inserts)
Synonym(s):
cell culture inserts, plate inserts, tissue culture dishes, tissue culture plates
Select a Size
€508.00
Select a Size
About This Item
€508.00
Recommended Products
Product Name
Corning® Netwell® inserts, membrane diam. 24 mm, membrane size 440 μm , mesh
material
clear
clear wells
polyester membrane
polyester plate
round wells
sterility
sterile
feature
lid
skirt
packaging
pkg of 48 (8 x 6 well inserts)
manufacturer/tradename
Corning 3480
membrane diam.
24 mm
membrane size
440 μm , mesh
size
6 wells , per insert
color
clear
Looking for similar products? Visit Product Comparison Guide
General description
- Immunocytochemical staining
- Explant culture
- 3-D tissue culture
- Coarse filtration of cell suspension
Features and Benefits
- Corning® Netwell® inserts have 440 μm polyester mesh bottoms attached to polystyrene to rings or housing.
- Supplies sterile and pre-loaded in 6-well microplates.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service