Skip to Content
Merck
All Photos(1)

Key Documents

CLLS1010

Sigma-Aldrich

SW48 Cells SMAD4 -/-

Human female colon, colorectal adenocarcinoma

Synonym(s):

CLL1008

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€4,060.00

€4,060.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 SET
€4,060.00

About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

€4,060.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

SW48 Cells SMAD4 -/-,

biological source

human female colon (Source Disease: Colorectal adenocarcinoma)

Quality Level

OMIM accession no.

storage temp.

−196°C

Gene Information

human ... SMAD4(4089)

General description

SW48 Cells SMAD4 -/- are colorectal adenocarcinoma, epithelial cells, from a human caucasian female (aged 82 years), with a ZFN knock out modification.
This product corresponds to ATCC Cat. No. CCL-231

Application

Media Renewal changes one to two times per week
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture Ratio: approx. 1:2 to 1:6
The base medium for this cell line is DMEM, Cat. No. D5671. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10%, Sodium pyruvate, Cat. No. S8636, at a final concentration of 1 mM, and L-glutamine, Cat. No. G7513, at a final concentration of 2mM.
The cell freezing medium is DMSO 1X (Cat. No. C6164)

Features and Benefits

These SW48 cells are adherent, with a doubling time of approx. 35 hours.
Zinc finger nuclease (ZFN) knock out on chromosome 18q21.1

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, PCR assay for cell line species confirmation.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Kit Components Also Available Separately

Product No.
Description
SDS

  • SW48 Cells SMAD4 -/- 1 vial

  • SW48 Control Cells 1 vial


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service