CLL1038
U2OS BFP-LMNB1 RFP-ACTB
human female bone (Disease source: Osteosarcoma)
About This Item
Recommended Products
Product Name
U2OS BFP-LMNB1 RFP-ACTB,
biological source
human female bone (Disease source: Osteosarcoma)
storage temp.
−196°C
Gene Information
human ... ACTB(60) , LMNB1(4001)
General description
U2OS BFP-LMNB1 RFP-ACTB are osteosarcoma, epithelial cells, from a human caucasian female (aged 15 years), with BFP-tagged and RFP-tagged modifications.
Application
Features and Benefits
U2OS cells are adherent, with a doubling time of approx. 29 hours.
Components
Quality
Preparation Note
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture Ratio: approx. 1:3-1:6
The base medium for this cell line is McCoy′s 5A1 Medium Modified, Cat. No. M9309. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration of 10%.
The cell freezing medium is DMSO 1X (Cat. No. C6164).
Legal Information
Disclaimer
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service