Skip to Content
Merck
All Photos(1)

Key Documents

SVCT-MI2

98031105, human breast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

SVCT-MI2, 98031105

biological source

human breast

shipped in

dry ice

storage temp.

−196°C

General description

For complete product information, please see SVCT-MI2

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service