Skip to Content
Merck
All Photos(1)

Documents

COR-L 23/CPR

96042336, human lung, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

COR-L 23/CPR, 96042336

biological source

human lung

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian lung, large cell carcinoma

Cell Line Description

The cell line COR-L23/CPR is a drug-resistant variant of COR-L23 (Sigma Catalogue number. 92031919). The line was developed by growing the parent line in increasing concentrations of cisplatin. Cisplatin accumulation is reduced in COR-L23/CPR and cross resistance to melphalan and other platinum compounds has been found. Cells tend to grow in clumps.

Application

Analysis of acquired drug resistance, study of large cell carcinoma of the lung

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 10,11
D16S539: 11,13
D5S818: 10,12
D7S820: 9
THO1: 9
TPOX: 8
vWA: 19

Culture Medium

RPMI 1640 + 2mM Glutamine + 0.05μg / ml Cisplatin + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:5 i.e. seeding at 2-5 x 10,000 cells / cm2 using 0.25 % trypsin or trypsin/EDTA; 5% CO2; 37°C. It is recommended to culture the cells without drug upon resuscitation until the first passage.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service