Skip to Content
Merck
All Photos(1)

Key Documents

SNU-C2B

96020932, human intestine, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

SNU-C2B, 96020932

biological source

human intestine

growth mode

Semi-adherent aggregates

karyotype

Pseudodiploid. Modal chromosome number is 46 at 70% with polyploidy at 16.4%. The del(3) (p23), t(4,?), t(8;?) (p22), del (9) (p13), del (11) (p13), t(1;8) (p13;p24), t(1;?) (q11,?) and 2 other marker chromosomes, if any, occur only sporadically.

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Colorectal Carcinoma

Cell Line Description

This cell line was derived from a fourth passage nude mouse xenograft of tumour fragments obtained from a 43 year old Asian female patient prior to therapy. The cells have a high concentration of receptors for EGF. This cell line has lost both chromosomes 8 and 11. This is one of 14 colorectal carcinoma cell lines derived by JG Park and associates during the years 1982 to 1985. These cells are tumourigenic in nude mice when 10E7 cells are inoculated subcutaneously.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS) + 1.0mM sodium pyruvate

Subculture Routine

Split sub-confluent cultures (70-80 %) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 37 C; 5% CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service