Skip to Content
Merck
All Photos(1)

Documents

BW1J

94101901, mouse skin, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BW1J, 94101901

biological source

mouse skin

growth mode

Adherent

karyotype

Hypotetraploid

morphology

Epithelial

products

Alpha-foetoprotein and Albumin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse hepatoma

Cell Line Description

A mouse hepatoma isolated from the BW7756 transplantable hepatoma carried in C57 leaden/J mice (see reference: Differentiation). Will synthesise in appreciable amounts only hepatic proteins that appear early in foetal development. The majority of cells contain at least two and often several nucleoli, fat droplets are rare. A mixture of BW1-J and Fao cells were treated with inactive Sendai Virus to produce hybrids at 1 hybrid colony/50 BW1-J cells.

Culture Medium

Coon′s modified Ham′s F12 (or Kaign′s modified Ham′s F12 + 45mg/L ascorbic acid + 18mg/L Inositol) + 2mM Glutamine + 5% Foetal Bovine Serum (FBS)

Subculture Routine

Split semi-confluent cultures (70%) 1:50 i.e. seeding at 2x1000 - 5x1000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C. Media change three times a week.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service