Skip to Content
Merck
All Photos(1)

Documents

LC-2/ad

94072247, human lung, Epithelial

Synonym(s):

LC-2/ad Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

LC-2/ad, 94072247

biological source

human lung

growth mode

Adherent

karyotype

Modal No 53-56

morphology

Epithelial

products

alpha-1-anti-trypsin

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human lung adenocarcinoma

Cell Line Description

Established from the pleural effusion of a pulmonary adenocarcinoma in a 51 year-old Japanese female. LC-2/ad cells exhibit an epithelial appearance and a tendency to form small domes. Immunocytochemically the cells are positive for CEA and cytokeratins, including cytokeratin No.18. LC-2/ad produces at least two functionally active trypsin inhibitors together with several proteinases in vitro.

Application

LC-2/ad has been used to study the ability of RXDX-105 as a therapy for cancers which are affected by RET (proto-oncogene tyrosine-protein kinase receptor) changes.

Culture Medium

RPMI 1640:HAMS F12 (1:1) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split confluent cultures 1:2-1:4 ie seeding at 2-5 x 10,000 cells/cm2 using 0.25% trypsin or trypsin/ EDTA ; 5%CO2; 37°C

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DNA, RNA, cDNA derived from ECACC mammalian cell lines allow screening for genes or expression patterns to identify lines most suitable for specific research.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service