Skip to Content
Merck
All Photos(1)

Documents

PC-3 Cell Line human

90112714, human prostate, Epithelial

Synonym(s):

PC.3 Cells, PC3 Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32190102

product name

PC-3 Cell Line human, 90112714, Caucasian prostate adenocarcinoma

biological source

human prostate

growth mode

Adherent

karyotype

2n = 46, triploid, modal no. 62

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human Caucasian prostate adenocarcinoma

Cell Line Description

Established from a grade 4 prostatic adenocarcinoma from a 62 year old male Caucasian. The cells grow in agar and produce tumours in nude mice. Exhibit low acid phosphatase and testosterone-5-α reductase activity. The Y chromosome could not be detected in this cell line by short tandem repeat (STR)-PCR analysis when tested at ECACC. It is a known phenomenon that due to the increased genetic instability of cancer cell lines the Y chromosome can be rearranged or lost resulting in lack of detection. The cell line is identical to the source provided by the depositor based on the STR-PCR analysis.

Application

PC-3 cell line has been used to study the antiproliferative effects of F. mughlea Pesmen on cancer cells. It has also been used to study the effects of palmitoylcarnitine (palcar) on the gene expression and pro-inflammatory pathways in prostate cancer.
Tumourigenicity studies

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11
D16S539: 11
D5S818: 13
D7S820: 8,11
THO1: 6,7
TPOX: 8,9
vWA: 17

Culture Medium

Coons Modified Ham’s F12 + 2mM Glutamine + 7% Foetal Bovine Serum (FBS).OR Kaign’s modified Ham’s F12 + 45mg/L ascorbic acid + 18 mg/L Inositol + 2mM Glutamine + 7% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:6 i.e. seeding at 2-5x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C. Medium change every 5 days. The initial subculture interval after cells are thawed may be longer than 7-9 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service