Skip to Content
Merck
All Photos(1)

Key Documents

3A sub E

90110508, human placenta, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

Product Name

3A sub E, 90110508

biological source

human placenta

growth mode

Adherent

karyotype

Not specified

morphology

Not specified

products

Human chorionic gonadotropin (hCG)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human placenta (post crisis); SV40 transformed

Cell Line Description

The cell line 3A sub E was derived from 3A(tPA-30-1)after the original line started to senesce and went through crisis. 3A subE has an unlimited lifespan and expresses human chorionic gonadotropin (hCG)

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 10,12
D13S317: 11
D16S539: 9,12
D5S818: 11,13
D7S820: 10
THO1: 9,9.3
TPOX: 8,11
vWA: 16,17

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Aspirate medium and add 0.25% trypsin/EDTA; incubate at room temperature until cells detach. Dilute with fresh medium and aliquot into new flasks at a split ratio 1:2. Renew fluid 1-2 times per week. Incubate culture at 37°C, 5% CO2 for initial 24 hour

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service