EIII
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
Human x Mouse Hybridoma hybrid
packaging
tube of 5 μg 86072804-DNA-5UG
pkg of vial of cells 86072804-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Fibroblast
products
Not specified
receptors
pJ3.11, p79a, CFTR DNA markers
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Human x mouse hybrid
Cell Line Description
This cell line may contain a sub-chromosomal fragment of human chromosome 17 bearing markers thought at the time to flank the cytotic fibrosis gene. The human DNA was donated from the human osteosarcoma cell line MNNG-HOS.
Application
Somatic cell hybrid mapping
Culture Medium
DMEM + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 37°C; 5% CO2.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service