Skip to Content
Merck
All Photos(1)

Documents

U266B1

85051003, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

U266B1, 85051003

biological source

human blood

growth mode

Suspension

karyotype

Not specified

morphology

Lymphoblast

products

IgE

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Human myeloma

Cell Line Description

This cell line was derived from peripheral blood of a patient with an IgE myeloma (epsilon2,lambda2). The line has been reported to secrete IgE and is used as fusion partner for the production of hybridomas. U266B1 has been reported to produce interleukin-6 (IL-6).

Application

Fusion partner

DNA Profile

STR-PCR Data: Amelogenin: X,Y
CSF1PO: 12,13
D13S317: 12
D16S539: 10
D5S818: 11,12
D7S820: 11,12
THO1: 5,7
TPOX: 8
vWA: 17

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Selection of the right fusion partner cell line is critical to obtain desired attributes of the hybridoma. Explore hybridoma fusion partners cell lines.

Related Content

Major technological advances have made the production of monoclonal antibodies quicker and more efficient. There are three established platforms for antibody discovery. We offer reagents for production of monoclonal antibody libraries using each of these techniques.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service