Skip to Content
Merck
All Photos(1)

Documents

HeLa-Mitotrap-Ap1g1-FKBP

15042203, human cervix, Epithelial, polygonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HeLa-Mitotrap-Ap1g1-FKBP, 15042203

biological source

human cervix

description

Human cervix carcinoma, knocksideways, rapid protein inactivation by rerouting to mitochondria for functional studies, adaptor protein (Ap1g1), genetically modified

growth mode

Adherent

morphology

Epithelial, polygonal

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Description

This cell line can be used to determine the function of a protein by rapidly inactivating the protein of interest by rapamycin induced rerouting of the protein to the mitochondria.

The HeLa-Mitotrap-Ap1g1-FKBP is a stably transfected HeLa cell line expressing two proteins containing rapamycin binding activity. To generate this cell line, the HeLa-Mitotrap cell line, ECACC 15042201, expressing a single construct expressing a mitochondrial trapping protein containing an FRB domain, (the term FRB refers to protein containing a FKBP12 and Rapamycin Binding (FRB) domains within the mTOR (mammalian Target of Rapamycin) protein, has been transfected with a second construct expressing siRNA-resistant subunits, FKBP-tagged subunits of the adaptor protein (AP1) complexes of clatharin-coated vesicles (CCVs) together with an FKBP and rapamycin-binding domain with a mitochondrial targeting signal. An FKBP domain, FKBP is a cellular chaperone protein and prolyl isomerase which is a target for rapamycin binding and will dimerize with FRB-containing protein. Upon treatment with rapamycin a target protein containing an FKBP domain, e.g, AP1, will dimerize with FRB domains and sequester the resulting protein complexes to the mitochondria. Treatment of cell lines containing both FRB and FKBP rapamycin binding proteins can sequester target proteins to the mitochondria within minutes of treatment rendering the cells ready for immediate assay.

The ?-FKBP construct was based on a pIRESneo2 plasmid encoding an siRNA-resistant form of a, which includes a brain-specific insert that acts as an epitope tag (Motley et al., 2006, PMID: 17035630). The FKBP coding sequence was amplified by PCR and inserted into the linker region of ?. The open reading frame was sequenced, the plasmid was transfected into HeLaM cells, and clonal cell lines were isolated as previously described (Motley et al., 2006).

Culture Medium

Dulbecco′s Modified Eagle′s Medium, high glucose (DMEM) + 2mM Glutamine + 10% Fetal Bovine Serum (FBS)

Subculture Routine

Trypsinise cultures at 80% confluence with 0.05% Trypsin/EDTA. Seed flasks at 2 x 104 cells/ cm2. Cultures must be incubated in a humidified 10% CO2/90% air incubator at 37oC.

Other Notes

Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service