Recommended Products
product name
FM-88, from human skin, 13012421
biological source
human skin
description
Human melanoma, metastasis
growth mode
Adherent
morphology
Fibroblastic
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−196°C
Cell Line Description
FM-88 (ESTDAB-029) was established from a metastatic cutaneous malignant melanoma of a male patient. The FM-88 cell line is one of more than 170 characterized melanoma cell lines collected during the ESTDAB project (European Searchable Tumour Line Database) part of the European Commission′s fifth framework infrastructures programme (contract no. QLRI-CT-2001-01325). ESTDAB contains information on HLA genotype and surface expression of HLA molecules, expression of tumour antigens, antigen processing capability, production of and response to cytokines and chemokines, apoptosis regulation and expression of adhesion/accessory molecules. This information is appended below. Such information allows researchers to identify cells possessing specific parameters important for studies of immunity, immunogenetics, gene expression, metastasis, response to chemotherapy, and other tumour biological experimentation.
DNA Profile
STR-PCR Data:
Amelogenin: X,Y
CSF1PO: 11
D13S317: 10,12
D16S539: 9,13
D5S818: 11
D7S820: 12
THO1: 7,9
TPOX: 9,11
vWA: 16,19
Amelogenin: X,Y
CSF1PO: 11
D13S317: 10,12
D16S539: 9,13
D5S818: 11
D7S820: 12
THO1: 7,9
TPOX: 9,11
vWA: 16,19
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Fetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) seed new cultures at 2-4 x10,000 cells/cm² using 0.05% trypsin or trypsin/EDTA; 5% CO2; 37°C. Saturation density 15 x 104 cells/cm2.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Cultures from PHE Culture Collections and supplied by Sigma are for research purposes only. Enquiries regarding the commercial use of a cell line are referred to the depositor of the cell line. Some cell lines have additional special release conditions such as the requirement for a material transfer agreement to be completed by the potential recipient prior to the supply of the cell line. Please view the Terms & Conditions of Supply for more information.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service