Skip to Content
Merck
All Photos(1)

Documents

CAD cell line from mouse

8100805, mouse nerve, Neuronal

Synonym(s):

CAD Cell Line

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CAD cell line from mouse, 08100805

biological source

mouse nerve

description

Mouse (B6/D2 F1 hybrid) catecholaminergic neuronal tumor

growth mode

Adherent

karyotype

Not specified

morphology

Neuronal

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Related Categories

Cell Line Origin

Mouse (B6/D2 F1 hybrid) catecholaminergic neuronal tumour

Cell Line Description

CAD cells (Cath.-a-differentiated) are a variant of a CNS catecholaminergic cell line established from a brain tumour in a transgenic mouse carrying wild-type SV40 T antigen under transcriptional control of rat tyrosine hydroxylase promoter. CAD has lost the immortalising oncogene and expresses neurone-specific proteins and synaptic vesicle proteins. CAD will differentiate to form neuronal processes in serum free conditions.

Application

Study of neuronal differentiation

Culture Medium

DMEM:HAMS F12 (1:1) + 2mM Glutamine + 8% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 seeding at 3 X 104 cells/cm2 Cells should be dislodged by gentle pipetting; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service