Skip to Content
Merck
All Photos(1)

Key Documents

108CC15

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Mouse x Rat Hybridoma nerve

packaging

tube of 5 μg 08062516-DNA-5UG
pkg of vial of cells 08062516-1VL

growth mode

Adherent

karyotype

Modal No. varied with passage see reference

morphology

Neuronal

products

Long processes after treatment with dibutyryl cAMP, dense core vesicles, excitable membranes, neurotransmitter enzymes, synthesis of neurohormones and uptake of catecholamines.

receptors

Receptors for neurohormones: delta opioid, PGE1 vasoactive intestinal polypeptide, adenosine, somatostatin and acetylcholine.

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse neuroblastoma x Rat glioma hybrid

Cell Line Description

The cell line108CC15 is formed by the fusion of mouse neuroblastoma N18TG2 and rat glioma C6-BU-1using inactivated Sendai virus. This cell line was renamed NG108-15 by Klee and Nirenberg. Sigma has two catalogue entries for the 108CC15 cell line which differ in that they were deposited by different groups: 108CC15 from the original group that derived the cell line (Sigma Catalogue number 08062516) and NG108-15 (Sigma Catalogue number 88112302). Several other cell lines in the Sigma catalogue have been derived from 108CC15. This is a cholinergic hybrid cell line.

Application

Used as a neuronal Model

Culture Medium

DMEM + 2 mM glutamine + 10% Foetal Bovine Serum (FBS) HAT (0.1 mM hypoxanthine, 10 μM aminopterin and 16 μM thymidine) Cells can be cultured without HAT provided they were cultured inbetween in HT medium for 3-4 days

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 1-2x10,000 cells/cm2 using 0.05% trypsin/EDTA; 5% CO2; 37°C. The cultures rapidly produce lactic acid through anaerobic glycolysis causing the media to become acidified. The medium must never be allowed to become acidic as cells detach in clumps and take many passages to recover. Frequent inspection of cultures is therefore required.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service