Skip to Content
Merck
All Photos(1)

Key Documents

CAS4104-EP

Caps for Eppendorf® S-4-104 & S-4x750 Rotors

aerosol-tight, for 5804/5804R & 5810/5810R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103913
Product CAS4104-EP is not currently sold in your country. Contact Technical Service

feature

aerosol-tight

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 022638937 (NA)
Eppendorf® 5820748001 (EU)

technique(s)

centrifugation: suitable

compatibility

for use with 5804 R
for use with 5804
for use with 5810 R
for use with 5810

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service