Skip to Content
Merck
All Photos(1)

Key Documents

CAA481-EP

Caps for Eppendorf® A-4-81 Rotor

aerosol-tight, for 5804/5804R & 5810/5810R

Synonym(s):

Lid for Eppendorf® FA-45-48-11 Rotor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103913
Product CAA481-EP is not currently sold in your country. Contact Technical Service

feature

aerosol-tight

packaging

pack of 2 ea

manufacturer/tradename

Eppendorf® 022638661 (NA)
Eppendorf® 5810724007 (EU)

technique(s)

centrifugation: suitable

compatibility

for use with 5804 R
for use with 5804
for use with 5810 R
for use with 5810

General description

  • Caps for Eppendorf® A-4-81 Rotor Lid, aerosol-tight, for 500 mL rectangular bucket.
  • Set of 2 pieces.
  • For centrifuge 5810/5810R and 5910 R swing bucket rotor.

Features and Benefits

  • Aerosol-tight caps effectively minimize air resistance within the rotor.
  • Suitable for 500 mL rectangular bucket
  • Ideal for centrifuge 5810/5810R and 5910 R swing bucket rotor.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service