Skip to Content
Merck
All Photos(1)

Documents

C9593

Sigma-Aldrich

Catechol-UL-14C

≥98% (HPLC), solid

Synonym(s):

pyrocatechol-ul-14C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(14C6H4-1,2-[OH]2
CAS Number:
Molecular Weight:
110.11
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

solid

extent of labeling

5-25 mCi per mmol

storage temp.

2-8°C

SMILES string

Oc1ccccc1O

Looking for similar products? Visit Product Comparison Guide

Packaging

Combi-vial

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service