Skip to Content
Merck
All Photos(1)

Key Documents

C8480

Sigma-Aldrich

D-erythro-Ceramine C8

98% (TLC), solid

Synonym(s):

N-Octyl-D-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€803.00

€803.00


Please contact Customer Service for Availability


Select a Size

Change View
25 MG
€803.00

About This Item

Empirical Formula (Hill Notation):
C26H53NO2
Molecular Weight:
411.70
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:
NACRES:
NA.77

€803.00


Please contact Customer Service for Availability

Quality Level

Assay

98% (TLC)

form

solid

color

white

solubility

DMSO: 25 mg/mL
ethanol: 25 mg/mL

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)NCCCCCCCC

InChI

1S/C26H53NO2/c1-3-5-7-9-11-12-13-14-15-16-17-18-20-22-26(29)25(24-28)27-23-21-19-10-8-6-4-2/h20,22,25-29H,3-19,21,23-24H2,1-2H3/b22-20+/t25-,26+/m0/s1

InChI key

CSRGRGSMEUKQSD-XQAXIDAFSA-N

Biochem/physiol Actions

Inert to ceramidases; induces peak DNA fragmentation in 6 hrs; induces apoptosis in U937 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Karasavvas et al.
European journal of biochemistry, 236(2), 729-737 (1996-03-01)
We investigated the ability of N-octanoyl-sphingosine (C8-Cer) stereoisomers, N-octanoyl-DL-erythro-dihydrosphingosine (DL-e-DHC8-Cer), and a new ceramide derivative, N-octyl-D-erythro-sphingosine (D-e-C8-Ceramine), to induce apoptosis in U937 cells. We found the C8-Cer stereoisomers to be stereospecific with the D- and L-threo stereoisomers being severalfold more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service