Skip to Content
Merck
All Photos(1)

Key Documents

C7255

Sigma-Aldrich

Cystamine dihydrochloride

Synonym(s):

2,2′-Diaminodiethyl disulfide dihydrochloride, 2,2′-Dithiobis(ethylamine) dihydrochloride, Decarboxycystine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2SSCH2CH2NH2 · 2HCl
CAS Number:
Molecular Weight:
225.20
Beilstein:
3616850
EC Number:
MDL number:
UNSPSC Code:
12352204

Assay

≥98%

mp

217-220 °C (dec.) (lit.)

SMILES string

Cl[H].Cl[H].NCCSSCCN

InChI

1S/C4H12N2S2.2ClH/c5-1-3-7-8-4-2-6;;/h1-6H2;2*1H

InChI key

YUFRRMZSSPQMOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasue Ishii-Osai et al.
Journal of dermatological science, 67(1), 51-60 (2012-05-25)
N-propionyl-4-S-cysteaminylphenol (NPr-4-S-CAP) is selectively incorporated into melanoma cells and degrades them. However, it remains unclear whether NPr-4-S-CAP can induce cell death associated with the induction of host immune responses and tumor suppression in vivo. To examine the molecular mechanism of
Shosuke Ito et al.
Biochemical pharmacology, 84(5), 646-653 (2012-06-26)
Metastatic melanoma is resistant to conventional therapies. N-propionyl-4-S-cysteaminylphenol (NPrCAP), an N-protected sulfur-amine analog of tyrosine, is a good substrate for tyrosinase and is selectively incorporated into melanoma cells, causing cytotoxicity in vitro and in vivo. We have recently shown that
Thorsten Overath et al.
Molecular & cellular proteomics : MCP, 11(8), 467-477 (2012-05-05)
The post-translational modification of proteins with O-GlcNAc is involved in various cellular processes including signal transduction, transcription, translation, and nuclear transport. This transient protein modification enables cells or tissues to adapt to nutrient conditions or stress. O-Glycosylation of the 26
Rosana A S Fonseca et al.
Sensors (Basel, Switzerland), 11(11), 10785-10797 (2012-02-22)
A piezoelectric immunosensor based on gold nanoparticles (AuNPs) co-immobilized on a dithiol-modified surface is proposed for detection of human cardiac troponin T (TnT). Anti-human troponin T (anti-TnT) antibodies were covalently immobilized on the nanostructured electrode surface by thiol-aldehyde linkages. In
Yongqin Lv et al.
Analytical chemistry, 84(20), 8457-8460 (2012-09-25)
A novel approach to porous polymer monoliths hypercrosslinked to obtain large surface areas and modified with zwitterionic functionalities through the attachment of gold nanoparticles in a layered architecture has been developed. The capillary columns were used for the separation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service