Skip to Content
Merck
All Photos(1)

Key Documents

C5804

Sigma-Aldrich

Calcium phosphate tribasic

suitable for insect cell culture

Synonym(s):

Calcium hydroxyphosphate, HAp, Hydroxylapatite, Tribasic calcium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Ca5(OH)(PO4)3]x
CAS Number:
Molecular Weight:
502.31
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

technique(s)

cell culture | insect: suitable

mp

1100 °C (lit.)

SMILES string

[Ca++].[Ca++].[Ca++].[Ca++].O[Ca+].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

comparable product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Lyon et al.
The Journal of bone and joint surgery. American volume, 95(23), 2088-2096 (2013-12-07)
Recombinant human bone morphogenetic protein-2 (rhBMP-2) applied on an absorbable collagen sponge improves open tibial fracture-healing as an adjunct to unreamed intramedullary nail fixation. We evaluated rhBMP-2 and a new, injectable calcium phosphate matrix (CPM) formulation in acute closed tibial
Tie Liu et al.
Bone, 56(1), 110-118 (2013-06-05)
As an alternative to an autologous bone graft, deproteinized bovine bone (DBB) is widely used in the clinical dentistry. Although DBB provides an osteoconductive scaffold, it is not capable of enhancing bone regeneration because it is not osteoinductive. In order
Xi Chen et al.
The Journal of biological chemistry, 289(16), 11162-11174 (2014-03-13)
Calcium phosphate precipitates (CPPs) form complexes with DNA, which enter cells via endocytosis. Under this condition CPPs induce autophagy via the canonic autophagy machinery. Here we showed that CPP-induced autophagy was also dependent on endocytosis as the process was significantly
Liam M Grover et al.
Biomaterials, 34(28), 6631-6637 (2013-06-12)
Pyrophosphate ions are both inhibitors of HA formation and substrates for phosphatase enzymes. Unlike polyphosphates their hydrolysis results simultaneously in the complete loss of mineral formation inhibition and a localised elevation in orthophosphate ion concentration. Despite recent advances in our
Layal Safa et al.
Biochimie, 103, 80-88 (2014-04-22)
Replication protein A (RPA) is a single-stranded DNA binding protein that plays an essential role in telomere maintenance. RPA binds to and unfolds G-quadruplex (G4) structures formed in telomeric DNA, thus facilitating lagging strand DNA replication and telomerase activity. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service