Skip to Content
Merck
All Photos(1)

Key Documents

C5374

Sigma-Aldrich

CB-DMB

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€255.00
25 MG
€775.00

€255.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€255.00
25 MG
€775.00

About This Item

Empirical Formula (Hill Notation):
C22H23Cl2N7O
CAS Number:
Molecular Weight:
472.37
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

€255.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

solid

solubility

DMSO: 20 mg/mL

storage temp.

2-8°C

SMILES string

Cc1ccc(CNC(=N)NC(=O)c2nc(Cl)c(NCc3ccc(Cl)cc3)nc2N)c(C)c1

InChI

1S/C22H23Cl2N7O/c1-12-3-6-15(13(2)9-12)11-28-22(26)31-21(32)17-19(25)30-20(18(24)29-17)27-10-14-4-7-16(23)8-5-14/h3-9H,10-11H2,1-2H3,(H3,25,27,30)(H3,26,28,31,32)

InChI key

MGLQFHOCPGELNL-UHFFFAOYSA-N

Biochem/physiol Actions

CB-DMB is a specific and potent inhibitor of the Na+-Ca2+exchanger (NCX).
CB-DMB is a specific and potent inhibitor of the Na+-Ca2+exchanger (NCX); pan-inhibitor of the NA+-Ca2+exchanger isoforms.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service