Skip to Content
Merck
All Photos(1)

Key Documents

C5172

Sigma-Aldrich

(±)-Chlorpheniramine maleate salt

Sigma Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19ClN2 · C4H4O4
CAS Number:
Molecular Weight:
390.86
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

Sigma Reference Standard

packaging

vial of 250 mg

mp

130-135 °C (lit.)

SMILES string

CN(C)CCC(C1=CC=C(Cl)C=C1)C2=NC=CC=C2.O=C(/C=C\C(O)=O)O

InChI

1S/C16H19ClN2.C4H4O4/c1-19(2)12-10-15(16-5-3-4-11-18-16)13-6-8-14(17)9-7-13;5-3(6)1-2-4(7)8/h3-9,11,15H,10,12H2,1-2H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

DBAKFASWICGISY-BTJKTKAUSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

H1 Histamine receptor antagonist.

Packaging

Supplied in amber screw-cap vials

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Alagarsamy et al.
European journal of medicinal chemistry, 43(11), 2331-2337 (2007-11-23)
A series of novel 1-substituted-4-(2-methylphenyl)-4H-[1,2,4]triazolo[4,3-a]quinazolin-5-ones were synthesized by the cyclization of 2-hydrazino-3-(2-methylphenyl)-3H-quinazolin-4-one with various one carbon donors. The starting material 2-hydrazino-3-(2-methylphenyl)-3H-quinazolin-4-one was synthesized from 2-methyl aniline by a novel innovative route. The title compounds were tested for their in vivo
Veerachamy Alagarsamy et al.
Bioorganic & medicinal chemistry letters, 15(7), 1877-1880 (2005-03-23)
A series of novel 1-substituted-4-phenyl-1,2,4-triazolo[4,3-a]quinazolin-5(4H)-ones 7 were synthesized by the cyclization of 2-hydrazino-3-phenylquinazolin-4(3H)-one 6 with various one carbon donors. The starting material 2-hydrazino-3-phenylquinazolin-4(3H)-one 6, was synthesized from aniline 1 by a novel innovative route. When tested for their in vivo
Eef L Theunissen et al.
British journal of clinical pharmacology, 61(1), 79-86 (2006-01-05)
Previous studies have demonstrated that the antihistamines mequitazine, cetirizine and dexchlorpheniramine produce mild sedation after single doses. It is unknown, however, whether acute sedation persists after repeated dosing. Therefore, this study assessed the effects of repeated dosing of these antihistamines
Syed Muhammad Mukarram Shah
BMC complementary and alternative medicine, 15, 299-299 (2015-09-01)
Teucrium stocksianum (T. stocksianum) is one of the important members of the genus Teucrium which contains numerous biologically active compounds. Traditionally, it is used for the treatment of fever, pain, as expectorant and blood purifier. Researchers are trying to discover
P van Ruitenbeek et al.
Journal of psychopharmacology (Oxford, England), 22(6), 663-672 (2008-01-23)
Results from recent animal studies suggest an important role for histamine in memory functioning. Histaminergic drugs might prove beneficial for people suffering from memory impairment. To determine if histamine is involved in memory functioning this study evaluates the effects of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service