Skip to Content
Merck
All Photos(2)

Key Documents

C4529

Sigma-Aldrich

Coproporphyrin I tetramethyl ester

≥90% purity (HPLC), powder

Synonym(s):

Tetramethyl 3,8,13,18-tetramethyl-21H,23H-porphine-2,7,12,17-tetrapropionate

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€163.00

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€163.00

About This Item

Empirical Formula (Hill Notation):
C40H46N4O8
CAS Number:
Molecular Weight:
710.82
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

€163.00


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

Coproporphyrin I tetramethyl ester, ≥90% (HPLC)

Assay

≥90% (HPLC)

form

powder

color

purple to brown

mp

252-254 °C (lit.)

solubility

chloroform: 10 mg/mL, dark red

ε (extinction coefficient)

1,900-2,500 at 400 nm in chloroform at 1% (actual value given on label)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

COC(=O)CCc1c(C)c2cc3[nH]c(cc4nc(cc5[nH]c(cc1n2)c(C)c5CCC(=O)OC)c(C)c4CCC(=O)OC)c(C)c3CCC(=O)OC

InChI

1S/C40H46N4O8/c1-21-25(9-13-37(45)49-5)33-18-30-23(3)27(11-15-39(47)51-7)35(43-30)20-32-24(4)28(12-16-40(48)52-8)36(44-32)19-31-22(2)26(10-14-38(46)50-6)34(42-31)17-29(21)41-33/h17-20,41,44H,9-16H2,1-8H3/b29-17-,30-18-,31-19-,32-20-,33-18-,34-17-,35-20-,36-19-

InChI key

OVYASYKFNWTMII-QEMHYRKZSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Enzymatically prepared

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service