Skip to Content
Merck
All Photos(1)

Documents

C4403

Sigma-Aldrich

Calcium Ionophore A23187 mixed calcium magnesium salt

Approximate 1:15 molar ratio Ca:Mg.

Synonym(s):

Calcimycin, Calcium Ionophore mixed calcium magnesium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H37N3O6
Molecular Weight:
523.62
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

storage temp.

2-8°C

SMILES string

[Mg++].[Ca++].CNc1ccc2oc(CC3OC4(CCC3C)OC(C(C)CC4C)C(C)C(=O)c5ccc[nH]5)nc2c1C(O)=O

InChI

1S/C29H37N3O6.Ca.Mg/c1-15-10-11-29(17(3)13-16(2)27(38-29)18(4)26(33)20-7-6-12-31-20)37-22(15)14-23-32-25-21(36-23)9-8-19(30-5)24(25)28(34)35;;/h6-9,12,15-18,22,27,30-31H,10-11,13-14H2,1-5H3,(H,34,35);;/q;2*+2

InChI key

HKVXGTQLKZNXPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ionophore highly selective for Ca2+. Potentiates responses to NMDA but not to quisqualate. In cell culture, stimulates nitric oxide production by calmodulin-dependent constitutive nitric oxide synthase.

Other Notes

Ca and Mg contents may vary; actual content given on label.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rubina Pal et al.
Current protocols in cytometry, 92(1), e69-e69 (2020-02-25)
Understanding how immune cells respond to external stimuli such as pathogens or drugs is a key component of biomedical research. Critical to the immune response are the expression of cell-surface receptors and the secretion of cytokines, which are tightly regulated
Fatima El-Assaad et al.
PLoS pathogens, 10(3), e1003839-e1003839 (2014-03-22)
In patients with cerebral malaria (CM), higher levels of cell-specific microparticles (MP) correlate with the presence of neurological symptoms. MP are submicron plasma membrane-derived vesicles that express antigens of their cell of origin and phosphatidylserine (PS) on their surface, facilitating
Reiya Taniguchi et al.
Nature communications, 6, 8545-8545 (2015-10-16)
In vertebrates, the iron exporter ferroportin releases Fe(2+) from cells into plasma, thereby maintaining iron homeostasis. The transport activity of ferroportin is suppressed by the peptide hormone hepcidin, which exhibits upregulated expression in chronic inflammation, causing iron-restrictive anaemia. However, due
Arthur Liesz et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(2), 583-598 (2015-01-16)
Acute brain lesions induce profound alterations of the peripheral immune response comprising the opposing phenomena of early immune activation and subsequent immunosuppression. The mechanisms underlying this brain-immune signaling are largely unknown. We used animal models for experimental brain ischemia as
Shan Li et al.
Molecular cell, 74(6), 1123-1137 (2019-05-06)
Abnormal processing of stressed replication forks by nucleases can cause fork collapse, genomic instability, and cell death. Despite its importance, it is poorly understood how the cell properly controls nucleases to prevent detrimental fork processing. Here, we report a signaling

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service