C-39415
Melanocyte Growth Medium
SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml
Synonym(s):
Recommended with C-24210
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
aqueous solution
packaging
pkg of 1 vial
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Melanocyte Growth Medium SupplementMix (basal medium sold separately)
To form a complete medium, it should be combined with the basal medium: C-24210
To form a complete medium, it should be combined with the basal medium: C-24210
Application
Melanocyte Growth Medium is a serum-free, PMA (Phorbol Myristate Acetate)-containing Medium and has been developed for the in vitro cultivation of juvenile melanocytes. Since PMA is a tumor promoting mitogen, it may interfere with some experimental approaches. In these cases, we recommend using the PMA-free Melanocyte Growth Medium M2 (C-24300). The Melanocyte Growth Medium is optimized for primary human cells, but can also be used for equine, porcine, murine, and rat melanocytes.Recommended for:Normal Human Epidermal Melanocytes (NHEM), juvenile foreskin
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials
Quality
All lots of PromoCell Melanocyte Growth Medium are subjected to comprehensive quality control tests using primary human melanocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested melanocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Preparation Note
Final supplement concentrations after addition to the basal medium:Bovine Pituitary Extract: 0.004 ml / mlBasic Fibroblast Growth Factor (recombinant human): 1 ng / mlInsulin (recombinant human): 5 µg / mlHydrocortisone: 0.5 µg / mlPhorbol Myristate Acetate: 10 ng / ml
Storage and Stability
Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Subculture Routine
Click here for more information.
recommended
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Primary Human Epidermal Melanocytes and optimized growth media support in vitro cultivation.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service