Skip to Content
Merck
All Photos(1)

Key Documents

C3275

Sigma-Aldrich

Cytodex® 3 microcarrier beads

>175 μm particle size (spherical), in 0.9% NaCl solution

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41121800
NACRES:
NB.22

material

dextran beads

sterility

non-sterile

particle size

>175 μm (spherical)

density

1.04 g/cm3 at 25 °C

Looking for similar products? Visit Product Comparison Guide

General description

Dextran beads coated with denatured porcine-skin collagen bound to surface.
The microcarrier of choice for cells that may be difficult to culture in vitro.

Microcarrier culture is a technique which makes possible the practical high yield culture of anchorage-dependent cells. Cytodex has been specifically developed for the culture of a wide range of animal cells in culture volumes ranging from a few ml to more than 6000 L. Using Cytodex in simple suspension culture systems provides yields of several million cells per mL.

Formed by chemically coupling a thin layer of denatured collagen to the cross-linked dextran matrix, this is the microcarrier of choice for cells that may be difficult to culture in vitro, and particularly for cells with an epithelial like morphology.

Application

Cytodex™3 microcarrier beads are suitable for:
  • the isolation, transportation, and storage of cells
  • cells that might be challenging to culture in vitro, especially cells with an epithelial morphology, sensitive, differentiated, or low in plating efficiency (e.g., endocrine cells, hepatocytes)
  • large-scale production of viruses, cells, and recombinant cell products (e.g.  hormones, nucleic acids, enzymes, interferon)
For cells (particularly those of epithelial origin) which are normally difficult to grow in culture.

Features and Benefits

  • Designed for culture of primary cells or those with an epithelioid morphology, especially cells with low plating-efficiency or differentiated or sensitive cell-types which are normally difficult to grow, e.g. hepatocytes, endocrine cells.
  • Contains a layer of acid-denatured porcine collagen (gelatin) on the surface of the microporous matrix.
  • Designed to yield harvested cells of the highest viability.
  • Transparent for easy microscopic examination of attached cells.

Preparation Note

Prior to use, swell beads in PBS (50 mL/g) and autoclave at 120°C.

Legal Information

Cytodex is a registered trademark of Cytiva

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David K Giles et al.
Microbes and infection, 10(14-15), 1494-1503 (2008-10-04)
Confinement of the obligate intracellular bacterium Chlamydia trachomatis to a membrane-bound vacuole, termed an inclusion, within infected epithelial cells neither prevents secretion of chlamydial antigens into the host cytosol nor protects chlamydiae from innate immune detection. However, the details leading
Kun Ding et al.
mAbs, 9(2), 269-284 (2016-12-09)
Exudative age-related macular degeneration (AMD) is the most common cause of moderate and severe vision loss in developed countries. Intraocular injections of vascular endothelial growth factor (VEGF or VEGF-A)-neutralizing proteins provide substantial benefit, but frequent, long-term injections are needed. In
Huanjiao Jenny Zhou et al.
Nature communications, 9(1), 3303-3303 (2018-08-19)
Regulation of VEGFR2 represents an important mechanism for the control of angiogenesis. VEGFR2 activity can be regulated by post-translational modifications such as ubiquitination and acetylation. However, whether VEGFR2 can be regulated by SUMOylation has not been investigated. Here we show
Jian Guang Xu et al.
Stem cell research & therapy, 8(1), 10-10 (2017-01-25)
Adequate vascularization is crucial for supplying nutrition and discharging metabolic waste in freshly transplanted tissue-engineered constructs. Obtaining the appropriate building blocks for vascular tissue engineering (i.e. endothelial and mural cells) is a challenging task for tissue neovascularization. Hence, we investigated
George Haddad et al.
Journal of the American Society of Nephrology : JASN, 32(2), 323-341 (2021-01-23)
Renal ischemia-reperfusion (I/R) injury is a major cause of AKI. Noncoding RNAs are intricately involved in the pathophysiology of this form of AKI. Transcription of hypoxia-induced, long noncoding RNA H19, which shows high embryonic expression and is silenced in adults

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service