Skip to Content
Merck
All Photos(1)

Key Documents

C3270

Sigma-Aldrich

Cephalosporin C zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H19N3O8SZn
CAS Number:
Molecular Weight:
478.79
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[H][C@]12SCC(COC(C)=O)=C3N1C(=O)[C@H]2NC(=O)CCC[C@H](N)C(=O)O[Zn]OC3=O

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Cephalosporin C zinc salt is used to study the effect of transpeptidase expression, binding, and inhibition on bacterial cell wall mucopeptide synthesis.
Cephalosporin C zinc salt is used to study the effect of transpeptidase expression, binding, and inhibition on bacterial cell wall mucopeptide synthesis. Cephalosporin C has been used to determine activity during stopped-flow Fuorescence spectrometric measurements of association . It is also used as a substrate to detect Cephalosporinase activity .

Biochem/physiol Actions

Cephalosporins disrupt the synthesis of the peptidoglycan layer of bacterial cell walls.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jihong Wang et al.
The Journal of biological chemistry, 284(1), 595-609 (2008-10-09)
In a previous study, we examined thermodynamic parameters for 20 alanine mutants in beta-lactamase inhibitory protein (BLIP) for binding to TEM-1 beta-lactamase. Here we have determined the structures of two thermodynamically distinctive complexes of BLIP mutants with TEM-1 beta-lactamase. The
Heritable mass conversion of a mutant penicillinase-negative culture of Bacillus cereus to a positive fully de-repressed state.
M R Pollock et al.
Journal of general microbiology, 59(3), 303-316 (1969-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service