Skip to Content
Merck
All Photos(1)

Key Documents

C3145

Sigma-Aldrich

Cellulose phosphate

fibers, 50-150 μm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
47101511
Pricing and availability is not currently available.

form

fibers

particle size

50-150 μm

capacity

~3.8 meq/g

Looking for similar products? Visit Product Comparison Guide

Application

Cellulose phosphate is used in protein chromatography, ion exchange chromatography and cation exchange media. Cellulose phosphate (CELLPHOS) has been studied as a collector for analytical preconcentration of traces of Cd(II), Cr(III), Cu(II) and Ni(II) from aqueous sample solution and used to purify and characterize acid phosphatase from approximately 3000-year-old human bones from archeological excavations.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histone octamer trans-transfer: a signature mechanism of ATP-dependent chromatin remodelling unravelled in wheat nuclear extract.
Raut, V.V., et al.
Annals of Botany, 108, 1234-1246 (2011)
Stefan Söllner et al.
Extremophiles : life under extreme conditions, 10(6), 629-634 (2006-08-10)
A restriction endonuclease activity from Sulfolobus islandicus REN2H1 was purified by phosphocellulose and cation exchange chromatography. The enzyme cuts DNA at the recognition site GCwGC as could be shown by restriction analysis of plasmids and short synthetic duplex DNA. The
Marcos T Oliveira et al.
Methods in molecular biology (Clifton, N.J.), 554, 37-58 (2009-06-11)
The mitochondrion is the eukaryotic organelle that carries out oxidative phosphorylation, fulfilling cellular requirements for ATP production. Disruption of mitochondrial energy metabolism can occur by genetic and biochemical mechanisms involving nuclear-encoded proteins that are required at the mitochondrial DNA replication
Preparation of microtubule protein and purified tubulin from bovine brain by cycles of assembly and disassembly and phosphocellulose chromatography.
Herbert P Miller et al.
Methods in cell biology, 95, 3-15 (2010-05-15)
Nikita Avvakumov et al.
The Journal of general virology, 83(Pt 3), 517-524 (2002-02-14)
Adenovirus type 5 E1A proteins interact with cellular regulators of transcription to reprogram gene expression in the infected or transformed cell. Although E1A also interacts with DNA directly in vitro, it is not clear how this relates to its function

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service