Skip to Content
Merck
All Photos(1)

Key Documents

C-28051

Sigma-Aldrich

Monocyte Attachment Medium

250 ml

Synonym(s):

Primary Cell Culture Media

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€143.00

€143.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€143.00

About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

€143.00


Please contact Customer Service for Availability

Request a Bulk Order

form

aqueous solution

packaging

pkg of 250 ml

technique(s)

cell culture | mammalian: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

Monocyte Attachment Medium (Ready-to-use)

Application

The Monocyte Attachment Medium allows for the efficient adherence selection of monocytes from freshly isolated human mononuclear cells while maintaining optimal cell health. Thus, the time-consuming and costly immunomagnetic purification of monocytes is dispensable prior to experiments requiring adherent monocytes as a starting material. With properly performed washing steps large numbers of 80-90% pure monocytes can be obtained. Typical applications are generation of dendritic cells / macrophages as well as phagocytosis assays.Recommended for:Human mononuclear cells (freshly isolated)Human monocytes (freshly isolated)

Quality

All lots of PromoCell Monocyte Attachment Medium are subjected to comprehensive quality control tests using primary human mononuclear cells from peripheral blood. Each lot of Monocyte Attachment Medium is checked for the efficient adherence selection of highly viable monocytes. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).

Preparation Note

The Medium is chemically defined/xeno-free and does not need further supplementation.

Storage and Stability

Store the Medium at 2 to 8°C in the dark immediately after arrival. Do not freeze the Medium. If stored properly, the product is stable until the expiry date stated on the label. After opening, the shelf life of the medium is 6 weeks at 2 to 8°C. For use, pre-warm only an aliquot of the medium and keep the remaining medium refrigerated at 2 to 8°C.

Subculture Routine

Click here for more information.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Explore over 350 PromoCell products on Sigma.com, including cryopreserved blood cells and cell culture media for various human cells.

Protocols

Generate mature human dendritic immune cells from PBMCs or CD34+ stem cell derived monocytes in 7 days in defined and serum-free dendritic cell culture media. Explore over 350 PromoCell products.

Generate phagocytic M1 and M2 macrophages from human PBMCs in 10 days in serum-free and xeno-free cell culture media. Explore over 350 PromoCell products.

Questions

  1. Could you tell me the formula of this medium (C-28051)?

    1 answer
    1. This item is a product of PromoCell. The manufacturer considers the formulation proprietary. However, this is a complete medium that includes all the growth factors and supplements necessary for providing optimal cell health and efficient adherence selection of monocytes from freshly isolated mononuclear cells. This medium does not contain antibiotics or antimycotics and is formulated for use in an incubator with an atmosphere of 5% CO2.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service