Skip to Content
Merck
All Photos(1)

Key Documents

C2048

Sigma-Aldrich

Closures for 25 mm culture tubes, disposable

red

Synonym(s):

closure cap, closure caps

Sign Into View Organizational & Contract Pricing

Select a Size

500 EA
€297.00

€297.00


Please contact Customer Service for Availability


Select a Size

Change View
500 EA
€297.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.22

€297.00


Please contact Customer Service for Availability

material

polypropylene

description

Autoclavable

color

red

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service