About This Item
Assay
≥95%
storage temp.
2-8°C
SMILES string
C[C@H]1C\C=C\[C@H]2[C@H](O)C(=C)[C@@H](C)[C@H]3[C@H](Cc4ccccc4)NC(=O)[C@@]23[C@H](O)\C=C\[C@@](C)(O)C1
InChI
1S/C28H37NO4/c1-17-9-8-12-21-25(31)19(3)18(2)24-22(15-20-10-6-5-7-11-20)29-26(32)28(21,24)23(30)13-14-27(4,33)16-17/h5-8,10-14,17-18,21-25,30-31,33H,3,9,15-16H2,1-2,4H3,(H,29,32)/b12-8+,14-13+/t17-,18+,21-,22-,23+,24-,25+,27+,28+/m0/s1
InChI key
UKQNIEMKORIOQM-JXYXERRISA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Oral - Carc. 2
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service