Skip to Content
Merck
All Photos(1)

Key Documents

C112000

Sigma-Aldrich

DMT-dC(tac) Phosphoramidite

Synonym(s):

DMT-dC(tac) amidite, N-[2-[4-(1,1-dimethylethyl)phenoxy]acetyl]-5′-O-[bis(4-methoxyphenyl)phenylmethyl]-2′-deoxycytidine, 3′-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Sign Into View Organizational & Contract Pricing

Select a Size

18 G
€951.00

€951.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
18 G
€951.00

About This Item

Empirical Formula (Hill Notation):
C51H62N5O9P
CAS Number:
Molecular Weight:
920.04
UNSPSC Code:
12352202
NACRES:
NA.51

€951.00


Please contact Customer Service for Availability

Request a Bulk Order

type

for DNA synthesis

product line

Proligo Reagents

Assay

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

λ

conforms (UV/VIS Identity)

nucleoside profile

base: deoxycytidine
base protecting group: TAC
2' protecting group: none
5' protecting group: DMT
deprotection: fast

storage temp.

-10 to -25°C

SMILES string

CC(C)N(C(C)C)P(OCCC#N)O[C@H]1C[C@@H](O[C@@H]1COC(c2ccccc2)(c3ccccc3)c4ccccc4)N5C=CC(NC(=O)COc6ccc(cc6)C(C)(C)C)=NC5=O

InChI

1S/C49H58N5O7P/c1-35(2)54(36(3)4)62(59-31-17-29-50)61-42-32-46(53-30-28-44(52-47(53)56)51-45(55)34-57-41-26-24-37(25-27-41)48(5,6)7)60-43(42)33-58-49(38-18-11-8-12-19-38,39-20-13-9-14-21-39)40-22-15-10-16-23-40/h8-16,18-28,30,35-36,42-43,46H,17,31-34H2,1-7H3,(H,51,52,55,56)/t42-,43+,46+,62?/m0/s1

InChI key

NHKNJNORWFDRBB-ROEGVEHFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DMT-dC(tac) Phosphoramidite is a TAC Protected Phosphoramidite. Substitution of standard protecting groups with the labile TAC (tert.-butylphenoxyacetyl) protecting group results in ultra-fast and easy deprotection under mild conditions, suitable for oligonucleotides with base labile monomers and reporters as well as in-situ synthesis schemes on glass surfaces.

Other Notes

  • The deprotection of oligonucleotide synthesis products with the AMAreagent is ultra-fast: complete deprotection requires 10 minutes at 65 °C
  • Side reactions at C-monomers through transamination are eliminated
  • Not compatible with some base-labile modified nucleosides
  • dC(tac)-phosphoramidite can directly substitute for dC(bz)-phosphoramidite
  • No change is required in the reagents commonly used for DNA synthesis.Acetonitrile is used to dissolve phosphoramidite. The standard aceticanhydride capping reagent can be employed.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service