Key Documents
C079300
Sigma-Aldrich
Empty Synthesis Column
0.2μmol (Colourless Body)
Select a Size
100 EA
€178.00
€178.00
Please contact Customer Service for Availability
Request a Bulk Order
Select a Size
Change View
100 EA
€178.00
About This Item
UNSPSC Code:
12352200
NACRES:
NA.51
€178.00
Please contact Customer Service for Availability
Request a Bulk Order
packaging
pkg of 1 × 100 ea
compatibility
configured for ABI
configured for PerkinElmer
General description
Empty Synthesis Columns are color coded and colorless empty column bodies with filters. C079300 empty synthesis column is a colorless body designed for ABI and Expedite Synthesizers.
Legal Information
ABI is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Expedite is a trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries
Choose from one of the most recent versions:
Certificates of Analysis (COA)