Skip to Content
Merck
All Photos(1)

Key Documents

C0627

Sigma-Aldrich

Z-Gly-Pro-Leu-Gly-Pro

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H39N5O8
CAS Number:
Molecular Weight:
573.64
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

form

solid

storage temp.

2-8°C

SMILES string

CC(C)CC(NC(=O)C1CCCN1C(=O)CNC(=O)OCc2ccccc2)C(=O)NCC(=O)N3CCCC3C(O)=O

InChI

1S/C28H39N5O8/c1-18(2)14-20(25(36)29-15-23(34)33-13-7-11-22(33)27(38)39)31-26(37)21-10-6-12-32(21)24(35)16-30-28(40)41-17-19-8-4-3-5-9-19/h3-5,8-9,18,20-22H,6-7,10-17H2,1-2H3,(H,29,36)(H,30,40)(H,31,37)(H,38,39)

InChI key

DGRDYZZPQWDRBB-UHFFFAOYSA-N

Substrates

A substrate for collagenase.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous analyses of clostridium collagenase and neutral proteinase activities with a single synthetic substrate.
K Nomura
Analytical biochemistry, 107(1), 96-102 (1980-09-01)
K Nomura et al.
Biochemistry, 30(25), 6115-6123 (1991-06-25)
The sea urchin hatching enzyme (HEz) is a protease capable of dissolving the fertilization envelope that surrounds the embryo as a protective coat during early development. We have now purified a 37-kDa active enzyme from the supernatant fluid of hatched
T Morita et al.
Journal of biochemistry, 82(5), 1495-1498 (1977-11-01)
Twenty peptide-4-methylcoumarin amides (MCA) were newly synthesized and tested as possible substrates for alpha-thrombin, factor Xa, kallikreins, urokinase, and plasmin. These fluorogenic peptides contained arginine-MCA as the carboxyl-terminus. Release of 7-amino-4-methylcoumarin was determined fluorometrically. Of these peptides, the following were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service