Skip to Content
Merck
All Photos(1)

Documents

C0619

Sigma-Aldrich

III-31-C

≥98% (HPLC), solid

Synonym(s):

N-[[[(2R,3S)-3-[[(1,1-dimethylethoxy)carbonyl]amino]-2-hydroxy-4-phenylbutyl](phenylmethyl)amino]carbonyl]-L-leucyl-L-valine methyl ester, WPE-III-31-C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H52N4O7
CAS Number:
Molecular Weight:
640.81
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

solid

storage condition

desiccated

solubility

DMSO: ≥9 mg/mL

storage temp.

−20°C

SMILES string

COC(=O)[C@@H](NC(=O)[C@H](CC(C)C)NC(=O)N(C[C@@H](O)[C@H](Cc1ccccc1)NC(=O)OC(C)(C)C)Cc2ccccc2)C(C)C

InChI

1S/C35H52N4O7/c1-23(2)19-28(31(41)38-30(24(3)4)32(42)45-8)36-33(43)39(21-26-17-13-10-14-18-26)22-29(40)27(20-25-15-11-9-12-16-25)37-34(44)46-35(5,6)7/h9-18,23-24,27-30,40H,19-22H2,1-8H3,(H,36,43)(H,37,44)(H,38,41)/t27-,28-,29+,30?/m0/s1

InChI key

DINAVFJXFRFCRE-BLFWRVEASA-N

Biochem/physiol Actions

III-31-C is a (hydroxyethyl)urea γ-secretase inhibitor that binds to γ-secretase at the substrate docking site. DAPT, also a γ-secretase inhibitor binds to the substrate docking site as well. III-31-C is useful for the affinity isolation and characterization of the protease complex and for testing other ihhibitors that affect the γ-secretase active site. The γ-secretase complex is composed of presenilin as a catalytic component and three integral membrane proteins, nicastrin, Aph-1 and Pen-2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service