Key Documents
BCS-576
CoolBox
XT MicroTube 24 Workstation
About This Item
UNSPSC Code:
41103000
NACRES:
NB.22
Pricing and availability is not currently available.
feature
Includes CoolBox XT System, CoolRack XT M24 (hold 24 x 1.5-2.0 mL microfuge tubes)
manufacturer/tradename
BioCision BCS-576
L × W × H
20 × 16 × 15 cm
color
green
General description
Easily transfer samples from the CoolBox XT cooling workstation to the robotic platform for brief cold processing (approx 20-30 minutes <10°C). CoolRack XT M24 tube module can be used to store samples in the refrigerator or freezer and then transferred to the CoolBox XT system for over 16 hours of ice-free cooling on the benchtop, in the tissue culture hood or in the clean room. Use the optional XT Freezing Core to keep the samples frozen for up to 8 hours or use with dry ice for snap freezing. Minimize risk of contamination from microorganisms and nucleases.
Applications include qPCR, RNA assays, tissue culture, snap-freeze bacteria, virus or protein samples, short transport of temperature-sensitive samples from lab to lab, field to lab, patient to clinic.
Legal Information
CoolBox is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)