Key Documents
BAF134810075
Bel-Art® Write-on Label Tape
roll size 15 yard, yellow, width 3/4 in., pk of 4
Synonym(s):
labelling tape, moisture resistant label tape, solvent resistant label tape
Select a Size
4 EA
€36.80
€36.80
Please contact Customer Service for Availability
Select a Size
Change View
4 EA
€36.80
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
€36.80
Please contact Customer Service for Availability
material
yellow
packaging
pk of 4
manufacturer/tradename
Bel-Art F13481-0075
roll size
15 yard
width
19.05 mm
3/4 in.
Related Categories
General description
Select from six tape colors that correspond with familiar hazard color codes to highlight safety information written on the labels. Use yellow for chemical reactivity, green for safety labels, blue indicates health hazards, red alerts to flammability; orange indicates a biohazard and white can describe a specific hazard. The tape is custom manufactured from proprietary materials that resist moisture and most solvents. Will adhere to PTFE fluoropolymer resin covered materials and any clean surface. A maximum temperature of 247 degrees C for 30 minutes can be endured.
Features and Benefits
  • No sticky residue lingers when tape is stripped off
  • Marks made with pencil, ball-point pen or solvent ink marker survive most laboratory conditions
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Write-on is a trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)